Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Sodium Hydroxide Solution (10N/Certified), Fisher Chemical™
Shop All Fisherbrand Products
Click to view available options
Quantity:
1 L
4 L
20 L
Packaging:
Poly Bottle
PolyPac
Specifications
Specifications
pH | 12.0 |
Boiling Point | >100°C |
Melting Point | -10°C |
Concentration | 9.95 to 10.05 |
UN Number | 1824 |
Density | 1.32g/mL |
Quantity | 20 L |
Grade | Certified |
Identification | Pass Test |
Packaging | PolyPac |
Show More |
Safety and Handling


DOTInformation | DOT Class 8, : Corrosive |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction