Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Sodium Hydroxide 10N Concentrate, Fisher Chemical
Shop All Fisherbrand Products

Click to view available options
Quantity:
100 mL
Packaging:
Poly Bottle
Description
- A volumetric standard solution, titrated at 25°C to pH 8.6 (phenolphthalein endpoint) against NIST™potassium acid phthalate.
- Each bottle contains exact amount of 10N concentrate required to make 1 liter of 1N or 10 liters of 0.1N (N/10) solution.
This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.
Specifications
Specifications
| pH | 12.0 |
| Boiling Point | >100°C |
| Melting Point | -10°C |
| Concentration | 10N |
| UN Number | 1824 |
| Density | 1.32g/cm³ |
| Quantity | 100 mL |
| Grade | Certified |
| Identification | Pass Test |
| Packaging | Poly Bottle |
| Show More |
Safety and Handling
| DOTInformation | Class 8: Corrosive |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction