Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Sodium Hydroxide Solution (0.1N (N/10)/Certified), Fisher Chemical™
Shop All Fisherbrand Products
Click to view available options
Quantity:
1 L
4 L
20 L
Packaging:
Poly Bottle
PolyPac™
Specifications
Specifications
| Concentration | 0.0995 to 0.1005N |
| UN Number | 1824 |
| Quantity | 20 L |
| Grade | Certified |
| Identification | Pass Test |
| Packaging | PolyPac™ |
| Traceability to NIST | LOT 84L Potassium acid phthalate |
| Physical Form | Liquid |
| Chemical Name or Material | Sodium Hydroxide Solution, 0.1N (N/10) |
Safety and Handling
| Hazard Statement | CAUTION! |
| Precautionary Statement | Emergency Overview May cause eye, skin, and respiratory tract irritation. Use personal protective equipment. Ensure adequate ventilation. Wash off immediately with plenty of water for at least 15 minutes. Get medical attention immediately if symptoms occur. Rinse immediately with plenty of water, also under the eyelids, for at least 15 minutes. Obtain medical attention. Move to fresh air. If breathing is difficult, give oxygen. Get medical attention immediately if symptoms occur. Do not induce vomiting. Obtain medical attention. Obtain medical attention. . NFPA Health:1 Flammability:0 Instability:0 |
| DOTInformation | DOT Class 8, : Corrosive |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction