Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Sodium Hydroxide Solution, 40%, Fisher Chemical™
Shop All Fisherbrand Products
Click to view available options
Quantity:
4 L
10 L
Packaging:
Poly Bottle
PolyPac™
Specifications
Specifications
pH | 12.0 |
Boiling Point | 141.7°C |
Melting Point | 5°C |
Quantity | 10 L |
Percent Purity | 40 to 42% |
Identification | Pass Test |
Packaging | PolyPac™ |
Vapor Pressure | 3mmHg at 37°C |
Physical Form | Liquid |
Chemical Name or Material | Sodium Hydroxide Solution, 40% (w/w) |
Safety and Handling


Hazard Statement | DANGER! |
Precautionary Statement | Emergency Overview Causes severe burns by all exposure routes. Wear personal protective equipment. Ensure adequate ventilation. Do not breathe vapors or spray mist. Do not get in eyes, on skin, or on clothing. Wash off immediately with plenty of water for at least 15 minutes. Immediate medical attention is required. Rinse immediately with plenty of water, also under the eyelids, for at least 15 minutes. Immediate medical attention is required. Move to fresh air. If breathing is difficult, give oxygen. Do not use mouth-to-mouth resuscitation if victim ingested or inhaled the substance; induce artifici. Do not use mouth-to-mouth resuscitation if victim ingested or inhaled the substance; induce artifici. Immediate medical attention is required. NFPA Health:3 Flammability:0 Instability:1 |
DOTInformation | DOT Class 8, : Corrosive |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction