Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Silver Nitrate Solution (0.1N (N/10)/Certified), Fisher Chemical™
Shop All Fisherbrand Products
Click to view available options
Quantity:
500 mL
1 L
4 L
20 L
Packaging:
Glass Bottle
PolyPac™
Specifications
Specifications
| Color | Colorless |
| UN Number | 3082 |
| Concentration | 0.0995 to 0.1005N |
| Grade | Certified |
| Identification | Pass Test |
| Packaging | Glass Bottle |
| Traceability to NIST | 999b POTASSIUM CHLORIDE |
| Vapor Pressure | 14mmHg at 20°C |
| Quantity | 4 L |
| Physical Form | Liquid |
| Show More |
Safety and Handling
| Hazard Category | Signal Word: CAUTION! |
| Hazard Statement | CAUTION! |
| Precautionary Statement | Emergency Overview May cause eye, skin, and respiratory tract irritation . The toxicological properties have not been fully investigated. Harmful to aquatic organisms, may cause long-term adverse effects in the aquatic environment. NFPA Health:1 Flammability:0 Instability:0 |
| DOTInformation | DOT Class 5.1, : Oxidizer |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction