Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
ChromaCare™ LC-MS Aqueous Rinse, Probe Wash 1, Thermo Scientific™
Shop All Fisherbrand Products
Click to view available options
Quantity:
2.5 L
Packaging:
Amber Glass Bottle
Description
- Suitable for cleaning sample preparation needle and drain lines on LC instrument after sampling
- Save time by using off-the-shelf blend — premade for convenience
- Helps to maintain clean sample lines and reduce sample carryover
- Packaged in 2.5L amber glass bottle
- Complementary to T001262500
Specifications
Specifications
| CAS | 75-05-8 |
| Percent Purity | 1.8 to 2.2% (Acetonitrile) |
| Concentration or Composition (by Analyte or Components) | Water with 2% Acetonitrile |
| Packaging | Amber Glass Bottle |
| Quantity | 2.5 L |
| Physical Form | Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction