Learn More
ChromaCare™ LC-MS Instrument Flush Solution
Description
- Designed by Thermo Scientific to maximize cleaning while preserving instrument integrity in Vanquish™ and UltiMate™ UHPLC systems with QQQ MS
- Comprised of a unique blend of polar protic and aprotic solvents for removing a wide variety of contaminants
- Particularly suitable for new LC/MS and UHPLC/MS installations to rapidly stabilize baseline
- A blend of 25% acetonitrile, 25% methanol, 25%, water, and 25% 2-propanol (IPA)
- Offered in 1L borosilicate bottles and 2.5L amber glass bottles
Specifications
Specifications
| Description | LC-MS Instrument Flush Solution |
| Packaging | Borosilicate glass bottle |
| CAS | 75-05-8; 67-56-1; 67-63-0; 7732-18-5 |
| Physical Form | Liquid |
| Color | Colorless |
| Quantity | 1 L |
| Percent Purity | 25% Acetonitrile, 25% Methanol, 25% Water, 25% 2-Propanol (IPA) |
| Grade | LC-MS |
RUO – Research Use Only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.