Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tris (Tromethamine) (Granular/USP), Fisher Chemical™

Catalog No. T393212 Shop All Fisherbrand Products
Click to view available options
Quantity:
500 g
2.5 kg
12 kg
Packaging:
Poly Bottle
Poly Pail

Chemical Identifiers

CAS 77-86-1
Molecular Formula C4H11NO3
Molecular Weight (g/mol) 121.136
MDL Number MFCD00004679
InChI Key LENZDBCJOHFCAS-UHFFFAOYSA-N
Synonym Trishydroxymethylaminomethane, Tris Buffer, Tris, THAM, Trizma™, 2-Amino-2-(hydroxymethyl)-1,3-propanediol, Tris base, Tris(hydroxymethyl)aminomethane, Trometamol
PubChem CID 6503
ChEBI CHEBI:9754
IUPAC Name 2-amino-2-(hydroxymethyl)propane-1,3-diol
SMILES C(C(CO)(CO)N)O

Specifications

Melting Point 168.5°C
Boiling Point 219°C
Color White
pH 10.4
Physical Form Powder or Solid
Quantity 2.5 kg
Identification Pass Test (USP)
Loss on Drying 1.0% max. (USP)
Linear Formula H2NC(CH2OH)3
Packaging Poly Bottle
Formula Weight 121.14
Percent Purity 99.0 to 101.0% (USP)
Grade USP
Chemical Name or Material Tris (Tromethamine)
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.