Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Urea (Certified ACS), Fisher Chemical™
Shop All Fisherbrand Products
Click to view available options
Quantity:
500 g
3 kg
50 kg
Packaging:
Fiber Drum
Poly Bottle
Specifications
Specifications
Decomposition Information | Decomposes information: >132°C |
Melting Point | 131°C |
Color | White |
Density | 1.335g/cm³ |
pH | 7.5 to 9.5 |
Ignition Residue | 0.01% max. |
Assay Percent Range | 99.0 to 100.5 % |
Packaging | Fiber Drum |
Linear Formula | NH2CONH2 |
Quantity | 50 kg |
Show More |
Safety and Handling

Hazard Category | Signal Word: CAUTION! |
Precautionary Statement | Emergency Overview May cause eye, skin, and respiratory tract irritation. NFPA Health:1 Flammability:0 Instability:0 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction