Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Xylenes (Certified ACS), Fisher Chemical™

Catalog No. X520 Shop All Fisherbrand Products
Click to view available options
Quantity:
500 mL
1 L
1 gal.
4 L
20 L
50 L
200 L
Packaging:
BasicPak
Glass Bottle
Poly Bottle
Safe-Cote™ Amber Glass 
Steel Drum
Steel Pail

Mixture of ortho, meta, and para isomers and may contain some Ethylbenzene

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1330-20-7, 100-41-4
MDL Number MFCD00077264

Specifications

Melting Point -34°C
Boiling Point 136°C
Quantity 20 L
Identification Pass Test
Packaging Steel Pail
Physical Form Liquid
Residue after Evaporation 0.002% max.
Assay Percent Range Assay: ≥98.5%
Molecular Formula C8H10
Vapor Pressure 8.29mmHg at 25°C
Water 0.05% max.
Synonym Xylol, Dimethylbenzene
Formula Weight 106.17
Percent Purity ≥98.5%
Grade Assay: ≥98.5%
Chemical Name or Material Xylenes
Show More Show Less
Hazard Statement WARNING!
Precautionary Statement Emergency Overview
Flammable liquid and vapor. Harmful if absorbed through skin or if inhaled. Causes eye, skin, and respiratory tract irritation. Inhalation may cause central nervous system effects. Aspiration hazard if swallowed - can enter lungs and cause damage. Use personal protective equipment. Keep away from open flames, hot surfaces and sources of ignition. Use only under a chemical fume hood. Wash off immediately with plenty of water for at least 15 minutes. Obtain medical attention. Rinse immediately with plenty of water, also under the eyelids, for at least 15 minutes. Obtain medical attention. Obtain medical attention. Move to fresh air.

NFPA

Health:2
Flammability:3
Instability:0
DOTInformation DOT Class 3, : Flammable Liquid
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.