Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ KF1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA571364
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA571364 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA571364 Supplier Invitrogen™ Supplier No. PA571364
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

KF1 Polyclonal Antibody for Western Blot

The protein encoded by this gene contains a RING-H2 finger, a motif known to be involved in protein-protein and protein-DNA interactions. This gene is highly expressed in normal cerebellum, but not in the cerebral cortex. The expression of the rat counterpart in the frontal cortex and hippocampus was shown to be induced by elctroconvulsive treatment (ECT) as well as chronic antidepressant treatment, suggesting that this gene may be a molecular target for ECT and antidepressants. [provided by RefSeq].
TRUSTED_SUSTAINABILITY

Specifications

Antigen KF1
Applications Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 2% sucrose and 0.09% sodium azide
Gene RNF103
Gene Accession No. O00237
Gene Alias Adrg34; E3 ubiquitin-protein ligase RNF103; hKF-1; KF1; KF-1; RING finger protein 103; RING-type E3 ubiquitin transferase RNF103; RNF103; Zfp103; Zfp-103; zinc finger protein 103; zinc finger protein 103 homolog; zinc finger protein expressed in cerebellum
Gene Symbols RNF103
Host Species Rabbit
Immunogen synthetic peptide directed towards the middle region of human KF1.
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 7844
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.