Learn More
Ansell KleenGuard™ KGA10 Lab Coats
GREENER_CHOICE
Catalog No. 17101157 Shop All Ansell Products
Change view
Click to view available options
Size:
Medium
Large
X-Large
2X-Large
3X-Large
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size
17-101-157 Medium
17-101-158 Large
17-101-159 X-Large
17-101-160 2X-Large
17-101-161 3X-Large
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
This item is not returnable. View return policy
Catalog No. 17-101-157 Supplier Ansell Supplier No. 67314
Add to Cart
Add to Cart
This item is not returnable. View return policy
Highly breathable spunbond fabric lab coats are CE Category I PPE and are free from natural rubber latex and silicone materials.
  • For non-hazardous particulate protection
  • Lightweight and highly breathable spunbond fabric allows moisture to escape for maximum comfort
  • 4-Snap Closure, Elastic Wrists, No Pockets
  • Not made with natural rubber latex or silicone
  • Recyclable with the RightCycle™ Program
TRUSTED_SUSTAINABILITY
Specifications
Product Type General Purpose Lab Coat
Size Medium
Gender Unisex
Color White
Closure Type Snaps
Cuff Style Elastic
Disposable Single-use
Sterility Non-sterile
No. of Pockets None
Latex Free Yes
Model KGA10
Seam Type Stitched