Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
KGF/FGF-7 Antibody (001), Janelia Fluor™ 646, Novus Biologicals™

Rabbit Monoclonal Antibody
Supplier: Novus Biologicals NBP290560JF646
Description
KGF/FGF-7 Monoclonal antibody specifically detects KGF/FGF-7 in Mouse samples. It is validated for ELISASpecifications
KGF/FGF-7 | |
Monoclonal | |
Janelia Fluor 646 | |
50mM Sodium Borate | |
Rabbit | |
Protein A purified | |
RUO | |
Primary | |
Mouse | |
Purified |
ELISA | |
001 | |
ELISA | |
FGF-7, fibroblast growth factor 7HBGF-7, Heparin-binding growth factor 7, keratinocyte growth factor, KGFfibroblast growth factor 7 (keratinocyte growth factor) | |
Rabbit immunized with purified, recombinant Mouse KGF/FGF-7 (Accession#: NP_032034.1, Cys32-Thr194) | |
0.1 mL | |
Angiogenesis, Biologically Active Proteins, Cell Cycle and Replication, Stem Cell Markers | |
2252 | |
Store at 4C in the dark. | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction