Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ KGF Monoclonal Antibody (02)
Mouse Monoclonal Antibody
Supplier: Invitrogen™ MA529290
Description
This product is preservative free. It is recommended to add sodium azide to avoid contamination (final concentration 0.05%-0.1%). This antibody has specificity for Human FGF7/FGF-7/KGF.
Plays an important role in the regulation of embryonic development, cell proliferation and cell differentiation. Required for normal branching morphogenesis. Growth factor active on keratinocytes. Possible major paracrine effector of normal epithelial cell proliferation.
Specifications
KGF | |
Monoclonal | |
1 mg/mL | |
PBS with no preservative | |
P21781 | |
FGF7 | |
Recombinant Human FGF7/FGF-7/KGF Protein (Cys32-Thr194). | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG1 |
ELISA | |
02 | |
Unconjugated | |
FGF7 | |
Fgf7; FGF-7; Fibroblast growth factor 7; HBGF-7; Heparin-binding growth factor 7; keratinocyte growth factor; Kgf | |
Mouse | |
Protein A | |
RUO | |
2252 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction