Learn More
KGF Polyclonal Antibody, PeproTech®, Invitrogen™
Rabbit Polyclonal Antibody
$250.00
Specifications
Antigen | KGF |
---|---|
Concentration | 0.1-1.0 mg/mL |
Applications | ELISA, Western Blot |
Classification | Polyclonal |
Conjugate | Unconjugated |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
500P1950UG
![]() |
Invitrogen™
500P1950UG |
50 μg |
Each of 1 for $250.00
|
|
|||||
Description
AA Sequence of recombinant protein: CNDMTPEQMA TNVNCSSPER HTRSYDYMEG GDIRVRRLFC RTQWYLRIDK RGKVKGTQEM KNNYNIMEIR TVAVGIVAIK GVESEFYLAM NKEGKLYAKK ECNEDCNFKE LILENHYNTY ASAKWTHNGG EMFVALNQKG IPVRGKKTKK EQKTAHFLPM AIT. Preparation: Produced from sera of rabbits immunized with highly pure Recombinant Human KGF (FGF-7). Anti-Human KGF (FGF-7)-specific antibody was purified by affinity chromatography employing an immobilized Human KGF (FGF-7) matrix. Sandwich ELISA: To detect Human KGF (FGF-7) by sandwich ELISA (using 100 μL/well antibody solution) a concentration of 0.5-2.0 μg/mL of this antibody is required. This antigen affinity purified antibody, in conjunction with PeproTech Biotinylated Anti-Human KGF (FGF-7) (500-P19Bt) as a detection antibody, allows the detection of at least 0.2-0.4 ng/well of Recombinant Human KGF (FGF-7). Western Blot: To detect Human KGF (FGF-7) by Western Blot analysis this antibody can be used at a concentration of 0.1-0.2 μg/mL. Used in conjunction with compatible secondary reagents the detection limit for Recombinant Human KGF (FGF-7) is 1.5-3.0 ng/lane, under either reducing or non-reducing conditions.
Plays an important role in the regulation of embryonic development, cell proliferation and cell differentiation. Required for normal branching morphogenesis. Growth factor active on keratinocytes. Possible major paracrine effector of normal epithelial cell proliferation.Specifications
KGF | |
ELISA, Western Blot | |
Unconjugated | |
Rabbit | |
Human | |
P21781 | |
2252 | |
E.coli-derived Recombinant Human KGF (FGF-7). | |
Antigen Affinity Chromatography | |
FGF7 |
0.1-1.0 mg/mL | |
Polyclonal | |
Lyophilized | |
RUO | |
PBS with no preservative | |
Fgf7; FGF-7; Fibroblast growth factor 7; HBGF-7; Heparin-binding growth factor 7; keratinocyte growth factor; Kgf | |
FGF7 | |
Primary | |
-20°C |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.