Learn More
Invitrogen™ KGF Recombinant Rabbit Monoclonal Antibody (4L2O8)

Description
Positive Samples: HeLa, A-549, Raji, Mouse lung, Mouse spleen, Mouse uterus, Rat lung, Rat spleen, Rat uterus Immunogen sequence: MHKWILTWIL PTLLYRSCFH IICLVGTISL ACNDMTPEQM ATNVNCSSPE RHTRSYDYME GGDIRVRRLF CRTQWYLRID KRGKVKGTQE MKNNYNIMEI.
Specifications
Specifications
| Antigen | KGF |
| Applications | ELISA, Western Blot |
| Classification | Recombinant Monoclonal |
| Clone | 4L2O8 |
| Concentration | 1.5 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 0.05% BSA, 50% glycerol and 0.02% sodium azide; pH 7.3 |
| Gene | FGF7 |
| Gene Accession No. | P21781, P36363, Q02195 |
| Gene Alias | Fgf7; FGF-7; Fibroblast growth factor 7; HBGF-7; Heparin-binding growth factor 7; keratinocyte growth factor; Kgf |
| Show More |
Safety and Handling
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.