Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tocris Bioscience™ KH 7
GREENER_CHOICE

Catalog No. 383410
Change view
Click to view available options
Quantity:
10 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
38-341-0 10 mg
38-345-0 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 38-341-0 Supplier Tocris Bioscience™ Supplier No. 3834/10
Add to Cart
Add to Cart
This item is not returnable. View return policy

Selective soluble adenylyl cyclase inhibitor

Selective soluble adenylyl cyclase (sAC) inhibitor (IC50 = 3 - 10 μM in vivo). Inert towards transmembrane adenylyl cyclase (tmAC) in vitro and in whole cells at concentrations up to 300 μM. Blocks synthesis of cAMP and displays an antiapoptotic effect at concentrations of 1 - 100 μM.

Chemical Identifiers

CAS 330676-02-3
Molecular Formula C17H15BrN4O2S

Specifications

Quantity 10 mg
Formula Weight 419.3
Purity >99%
Molecular Weight (g/mol) 419.297
Chemical Name or Material KH 7
Recommended Storage Store at +4°C
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.