Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
KHDC1 Antibody, DyLight 594, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP306228DL594
Description
KHDC1 Polyclonal specifically detects KHDC1 in Human samples. It is validated for Immunocytochemistry/Immunofluorescence.Specifications
KHDC1 | |
Polyclonal | |
Immunocytochemistry/Immunofluorescence | |
bA257K9.4, C6orf148, chromosome 6 open reading frame 148, KH homology domain containing 1, KH homology domain-containing protein 1, MGC10818, NDG1 | |
Produced in rabbits immunized with E. coli-derived Human KHDC1 fragment. | |
0.1 mL | |
Primary | |
Human | |
Purified |
Immunofluorescence | |
DyLight 594 | |
50mM Sodium Borate | |
Rabbit | |
Antigen and protein A Affinity-purified | |
RUO | |
80759 | |
Store at 4°C in the dark. | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction