Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
KHDC1 Monoclonal Antibody (OTI2G6), TrueMAB™, OriGene

Mouse Monoclonal Antibody
Supplier: OriGene TA810622

Description
KHDC1 family members are K-homology (KH) domain-containing RNA binding proteins that are involved in various aspects of RNA metabolism, ranging from transcription to RNA splicing, transportation, translation, and stability. KHDC1 is highly expressed in oocytes and interacts with cytoplasmic polyadenylation element-binding protein 1 (CPEB1), a key translational regulator controlling the polyA length of mRNAs. KHDC1 functions as a global translational repressor and induces apoptosis through an ER-dependent signaling pathway and that the C-terminal putative trans-membrane motif (TMM) is critical for its activities.Specifications
KHDC1 | |
Monoclonal | |
Unconjugated | |
KHDC1 | |
C6orf147, C6orf148, Em:AC019205.8, NDG1, bA257K9.4 | |
Mouse | |
Affinity Chromatography | |
RUO | |
80759 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |
Western Blot | |
OTI2G6 | |
PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
Q4VXA5 | |
KHDC1 | |
Full length human recombit protein of human KHDC1 produced in E.coli. | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG2a |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction