Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
KHDRBS2 Antibody (7G8C10) - BSA Free, Novus Biologicals™

Mouse Monoclonal Antibody
$548.00
Specifications
Antigen | KHDRBS2 |
---|---|
Clone | 7G8C10 |
Concentration | 1 mg/mL |
Dilution | Western Blot 1:500 - 1:2000, Flow Cytometry 1:200 - 1:400, ELISA 1:10000, Immunohistochemistry 1:200 - 1:1000, Immunohistochemistry-Paraffin 1:200 - 1:1000, CyTOF-ready |
Applications | Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin), CyTOF |
Description
KHDRBS2 Monoclonal specifically detects KHDRBS2 in Human, Mouse samples. It is validated for Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Immunohistochemistry-Paraffin, CyTOF-ready.Specifications
KHDRBS2 | |
1 mg/mL | |
Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin), CyTOF | |
Unconjugated | |
RUO | |
PBS with 0.05% Sodium Azide | |
202559 | |
Purified recombinant fragment of human KHDRBS2 (AA: 160-349) expressed in E. Coli. | |
Primary | |
39 kDa |
7G8C10 | |
Western Blot 1:500 - 1:2000, Flow Cytometry 1:200 - 1:400, ELISA 1:10000, Immunohistochemistry 1:200 - 1:1000, Immunohistochemistry-Paraffin 1:200 - 1:1000, CyTOF-ready | |
Monoclonal | |
Mouse | |
Human, Mouse | |
hSLM-1, KH domain containing, RNA binding, signal transduction associated 2, KH domain-containing, RNA-binding, signal transduction-associated protein 2, MGC26664, Sam68-like mammalian protein 1, SLM1bA535F17.1, SLM-1FLJ38664 | |
KHDRBS2 | |
IgG1 | |
Affinity Purified |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title