Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

KHDRBS2 Mouse anti-Human, Mouse, Clone: 7G8C10, Abnova™

Catalog No. 89934402 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-934-402 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-934-402 Supplier Abnova Corporation Supplier No. MAB12249
Add to Cart
Add to Cart

Mouse monoclonal antibody raised against partial recombinant human KHDRBS2.

Specifications

Antigen KHDRBS2
Applications ELISA, Flow Cytometry, Immunohistochemistry (PFA fixed), Western Blot
Classification Monoclonal
Clone 7G8C10
Conjugate Unconjugated
Description Mouse monoclonal antibody raised against partial recombinant human KHDRBS2.
Dilution ELISA (1:10000) Flow Cytometry (1:200-1:400) Immunohistochemistry (1:200-1:1000) Western Blot (1:500-1:2000) The optimal working dilution should be determined by the end user.
Formulation In PBS (0.05% sodium azide)
Gene Alias FLJ38664/MGC26664/SLM-1/SLM1/bA535F17.1
Gene Symbols KHDRBS2
Host Species Mouse
Immunogen Recombinant protein corresponding to amino acids 160-349 of human KHDRBS2.
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 202559
Target Species Human, Mouse
Content And Storage Store at 4°C. For long term storage store at -20°C.
Aliquot to avoid repeated freezing and thawing.
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.