Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ KHDRBS2 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA556127
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA556127 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA556127 Supplier Invitrogen™ Supplier No. PA556127
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: PDYNDEIRQE QLRELSYLNG SEDSGRGRGI RGRGIRIAPT APSRGRGGAI PPPPPPGRGV LTPRGSTVTR GA Highest antigen sequence identity to the following orthologs: Mouse - 94%, Rat - 94%.

RNA-binding protein that plays a role in the regulation of alternative splicing and influences mRNA splice site selection and exon inclusion. Binds both poly(A) and poly(U) homopolymers. Phosphorylation by PTK6 inhibits its RNA-binding ability. Induces an increased concentration-dependent incorporation of exon in CD44 pre-mRNA by direct binding to purine-rich exonic enhancer. Can regulate alternative splicing of NRXN1 in the laminin G-like domain 6 containing the evolutionary conserved neurexin alternative spliced segment 4 (AS4) involved in neurexin selective targeting to postsynaptic partners. Regulates cell-type specific alternative splicing of NRXN1 at AS4 and acts synergistically with SAM68 in exon skipping. In contrast acts antagonistically with SAM68 in NRXN3 exon skipping at AS4. Its phosphorylation by FYN inhibits its ability to regulate splice site selection. May function as an adapter protein for Src kinases during mitosis.
TRUSTED_SUSTAINABILITY

Specifications

Antigen KHDRBS2
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 0.7 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene KHDRBS2
Gene Accession No. Q5VWX1
Gene Alias 6330586C16Rik; hSLM-1; KH domain containing, RNA binding, signal transduction associated 2; KH domain-containing, RNA-binding, signal transduction-associated protein 2; KH RNA binding domain containing, signal transduction associated 2; Khdrbs2; mSLM-1; rSLM-1; Sam68-like mammalian protein 1; Slim1; SLM1; SLM-1
Gene Symbols KHDRBS2
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human KHDRBS2. Recombinant protein control fragment (Product #RP-94677).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 202559
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.