Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ KHDRBS3 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA564874
Description
Immunogen sequence: ADVPVVRGKPT LRTRGVPAPA ITRGRGGVTA RPVGVVVPRG TPTPRGVLST RGPVSRGRGL LTPRARGVPP TGYRPPPPPP TQETYGEYDY DDGYGTAYDE QSYDS Highest antigen sequence identity to the following orthologs - mouse 95%, rat 93%.
As a RNA-binding protein, KHDRBS3 plays a role in the regulation of alternative splicing and influences mRNA splice site selection and exon inclusion. KHDRBS3 may play a role as a negative regulator of cell growth. It inhibits cell proliferation and involved in splice site selection of vascular endothelial growth factor. It induces an increased concentration-dependent incorporation of exon in CD44 pre-mRNA by direct binding to purine-rich exonic enhancer. RNA-binding abilities are down-regulated by tyrosine kinase PTK6. It also involved in post-transcriptional regulation of HIV-1 gene expression.
Specifications
KHDRBS3 | |
Polyclonal | |
Unconjugated | |
Khdrbs3 | |
Etle; etoile; etoile, Sam68-like protein SLM-2; KH domain containing, RNA binding, signal transduction associated 3; KH domain-containing, RNA-binding, signal transduction-associated protein 3; KH RNA binding domain containing, signal transduction associated 3; KHDRBS3; RNA-binding protein Etoile; RNA-binding protein T-Star; rSLM-2; SALP; Sam68-like mammalian protein 2; Sam68-like phosphotyrosine protein; Sam68-like phosphotyrosine protein, T-STAR; SLM2; SLM-2; TSTAR; T-STAR | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
10656 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Western Blot, Immunocytochemistry | |
0.1 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
O75525 | |
Khdrbs3 | |
Recombinant Human KHDRBS3. | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction