Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
KHK Monoclonal Antibody (OTI4B8), TrueMAB™, OriGene

Mouse Monoclonal Antibody
Supplier: OriGene TA501283

Description
This gene encodes ketohexokinase that catalyzes conversion of fructose to fructose-1-phosphate. The product of this gene is the first enzyme with a specialized pathway that catabolizes dietary fructose. Alternatively spliced transcript variants encoding different isoforms have been identified.Specifications
KHK | |
Monoclonal | |
0.44 mg/mL | |
PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
P50053 | |
KHK | |
Full length human recombit protein of human KHK produced in HEK293T cell. | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG1 |
Flow Cytometry, Immunocytochemistry, Immunofluorescence, Western Blot | |
OTI4B8 | |
Unconjugated | |
KHK | |
ketohexokinase | |
Mouse | |
Affinity Chromatography | |
RUO | |
3795 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction