Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ KHK Monoclonal Antibody (OTI4E9)
GREENER_CHOICE

Catalog No. PIMA525137
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA525137 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA525137 Supplier Invitrogen™ Supplier No. MA525137
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

KHK Monoclonal Antibody for Western Blot, ICC/IF, IHC (P), Flow

KHK encodes the gene ketohexokinase that catalyzes conversion of fructose to fructose-1-phosphate. The splice variant presented encodes the highly active form found in liver. renal cortex. and small intestine. while the alternate variant encodes the lowe.
TRUSTED_SUSTAINABILITY

Specifications

Antigen KHK
Applications Flow Cytometry, Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Monoclonal
Clone OTI4E9
Concentration 0.66 mg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.02% sodium azide; pH 7.3
Gene KHK
Gene Accession No. P50053
Gene Alias fructokinase; hepatic fructokinase; KETHPRO; ketohexokinase; ketohexokinase (fructokinase); ketohexokinase isoform a; Khk; testicular tissue protein Li 102
Gene Symbols KHK
Host Species Mouse
Immunogen Full length human recombinant protein of KHK produced in HEK293T cell.
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 3795
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.