Learn More
KHK Mouse anti-Human, Clone: OTI1F1, lyophilized, TrueMAB™

Mouse Monoclonal Antibody
Supplier: OriGene CF501285

Description
Reconstitute with PBS (pH 7.3) and recommend to perform another round of desalting process using Product No. 7KMWCO
This gene encodes ketohexokinase that catalyzes conversion of fructose to fructose-1-phosphate. The product of this gene is the first enzyme with a specialized pathway that catabolizes dietary fructose. Alternatively spliced transcript variants encoding different isoforms have been identified.Specifications
KHK | |
Monoclonal | |
Unconjugated | |
KHK | |
ketohexokinase | |
Mouse | |
Affinity Chromatography | |
RUO | |
3795 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Lyophilized |
Flow Cytometry, Immunocytochemistry, Immunofluorescence, Western Blot | |
OTI1F1 | |
PBS with 8% trehalose and no preservative; pH 7.3 | |
P50053 | |
KHK | |
Full length human recombit protein of human KHK produced in HEK293T cell. | |
100 μg | |
Primary | |
Human | |
Antibody | |
IgG1 |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.