Learn More
Invitrogen™ KHK Polyclonal Antibody

Description
Immunogen sequence: YGDVVFVSKD VAKHLGFQSA EEALRGLYGR VRKGAVLVCA WAEEGADALG PDGKLLHSDA FPPPRVVDTL GAGDTFNASV IFSLSQGRSV QEALRFGCQV Highest antigen sequence indentity to the following orthologs: Rat - 88%, Mouse - 91%.
Specifications
Specifications
| Antigen | KHK |
| Applications | Immunohistochemistry (Paraffin), Western Blot |
| Classification | Polyclonal |
| Concentration | 0.1 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 |
| Gene | KHK |
| Gene Accession No. | P50053, P97328, Q02974 |
| Gene Alias | fructokinase; hepatic fructokinase; KETHPRO; ketohexokinase; ketohexokinase (fructokinase); ketohexokinase isoform a; Khk; testicular tissue protein Li 102 |
| Gene Symbols | KHK |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.