Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ KHK Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA596677
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA596677 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA596677 Supplier Invitrogen™ Supplier No. PA596677
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: MEEKQILCVG LVVLDVISLV DKYPKEDSEI RCLSQRWQRG GNASNSCTVL SLLGAPCAFM GSMAPGHVAD FVLDDLRRYS VDLRYTVFQT TGSVPIATVI INEASGSRTI LYYDRSLPDV SATDFEKVDL TQFKWIHIEG RNASEQVKML QRIDAHNTRQ PPEQKIRVSV EVEKPREELF QLFGYGDVVF VSKDVAKHLG FQSAEEALRG LYGRVRKGAV LVCAWAEEGA DALGPDGKLL HSDAFPPPRV VDTLGAGDTF NASVIFSLSQ GRSVQEALRF GCQVAGKKCG LQGFDGIV; Positive Samples: Mouse kidney, Mouse liver, Rat kidney.

KHK encodes the gene ketohexokinase that catalyzes conversion of fructose to fructose-1-phosphate. The splice variant presented encodes the highly active form found in liver. renal cortex. and small intestine. while the alternate variant encodes the lowe.
TRUSTED_SUSTAINABILITY

Specifications

Antigen KHK
Applications ELISA, Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 3.6 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Gene KHK
Gene Accession No. P50053, P97328, Q02974
Gene Alias fructokinase; hepatic fructokinase; KETHPRO; ketohexokinase; ketohexokinase (fructokinase); ketohexokinase isoform a; Khk; testicular tissue protein Li 102
Gene Symbols KHK
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 1-298 of human KHK (NP_0002121).
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 16548, 25659, 3795
Target Species Human, Mouse, Rat
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.