Learn More
Promega KHS1 Kinase Enzyme System, 10microg
Catalog No. PRV4108 Shop All Promega Corporation Products
Encompass
Change view
Click to view available options
Quantity:
10 μg
Catalog No. Quantity
PRV4108 10 μg
1 options
Catalog No. PRV4108 Supplier Promega Supplier No. V4108
Recombinant full-length human KHS1 was expressed by baculovirus in Sf9 insect cells using an N-terminal GST tag. KHS1 belongs to the ser/thr kinase family that has a STE20-like protein kinase domain, which stimulates the SAPK pathway.
  • Easily Screen and Profile KHS1 Kinase Inhibitors
  • Includes kinase, substrate and reaction buffer
  • Use with ADP−Glo™ Assay for bioluminescent detection of kinase activity
Specifications
Quantity 10 μg
Content And Storage <-65°C