Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ KHSRP Monoclonal Antibody (4F10D2)
GREENER_CHOICE

Catalog No. PIMA549240
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA549240 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA549240 Supplier Invitrogen™ Supplier No. MA549240
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Adding 0.2 mL of distilled water will yield a concentration of 500 μg/mL. Positive Control - WB: human Hela whole cell, human MCF-7 whole cell. ICC/IF: MCF-7 cell. Flow: U251 cell. Store at -20°C for one year from date of receipt. After reconstitution, at 4°C for one month. It can also be aliquotted and stored frozen at -20°C for six months. Avoid repeated freeze-thaw cycles.

The KHSRP gene encodes a multifunctional RNA-binding protein implicated in a variety of cellular processes, including transcription, alternative pre-mRNA splicing, and mRNA localization.
TRUSTED_SUSTAINABILITY

Specifications

Antigen KHSRP
Applications Flow Cytometry, Western Blot, Immunocytochemistry
Classification Monoclonal
Clone 4F10D2
Concentration 500 μg/mL
Conjugate Unconjugated
Formulation PBS with 4mg trehalose and no preservative
Gene KHSRP
Gene Accession No. Q92945
Gene Alias 6330409F21Rik; Far upstream element-binding protein 2; FBP2; Fubp2; FUSE binding protein 2; FUSE-binding protein 2; KH type-splicing regulatory protein; KHE3; KHSRP; KH-type splicing regulatory protein; KH-type splicing regulatory protein (FUSE binding protein 2); KSRP; MAP2 RNA trans-acting protein 1; MAP2 RNA trans-acting protein MARTA1; MARTA1; p75
Gene Symbols KHSRP
Host Species Mouse
Immunogen E.coli-derived human KHSRP recombinant protein (Position: D116-R478).
Purification Method Antigen affinity chromatography
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 8570
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Lyophilized
Isotype IgG2b
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.