Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ KHSRP Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA5103840
Description
Antibody detects endogenous levels of total KHSRP.
The KHSRP gene encodes a multifunctional RNA-binding protein implicated in a variety of cellular processes, including transcription, alternative pre-mRNA splicing, and mRNA localization.
Specifications
KHSRP | |
Polyclonal | |
Unconjugated | |
KHSRP | |
6330409F21Rik; Far upstream element-binding protein 2; FBP2; Fubp2; FUSE binding protein 2; FUSE-binding protein 2; KH type-splicing regulatory protein; KHE3; KHSRP; KH-type splicing regulatory protein; KH-type splicing regulatory protein (FUSE binding protein 2); KSRP; MAP2 RNA trans-acting protein 1; MAP2 RNA trans-acting protein MARTA1; MARTA1; p75 | |
Rabbit | |
Affinity Chromatography | |
RUO | |
16549, 171137, 8570 | |
-20°C | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
1 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
Q3U0V1, Q92945, Q99PF5 | |
KHSRP | |
A synthesized peptide derived from human KHSRP(Accession Q92945), corresponding to amino acid residues I84-L134. | |
100 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction