Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ KHSRP Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA578256
Description
Positive Control: A431, HeLa, mouse brain Predicted Reactivity: Chicken (90%) Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
The KHSRP gene encodes a multifunctional RNA-binding protein implicated in a variety of cellular processes, including transcription, alternative pre-mRNA splicing, and mRNA localization.
Specifications
KHSRP | |
Polyclonal | |
Unconjugated | |
KHSRP | |
6330409F21Rik; Far upstream element-binding protein 2; FBP2; Fubp2; FUSE binding protein 2; FUSE-binding protein 2; KH type-splicing regulatory protein; KHE3; KHSRP; KH-type splicing regulatory protein; KH-type splicing regulatory protein (FUSE binding protein 2); KSRP; MAP2 RNA trans-acting protein 1; MAP2 RNA trans-acting protein MARTA1; MARTA1; p75 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
16549, 171137, 8570 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
0.76 mg/mL | |
PBS with 20% glycerol and 0.025% ProClin 300; pH 7 | |
Q3U0V1, Q92945, Q99PF5 | |
KHSRP | |
Recombinant protein encompassing a sequence within the center region of human KHSRP. The exact sequence is proprietary. | |
100 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction