Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tocris Bioscience™ Ki 20227
GREENER_CHOICESDP

Catalog No. 448110 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
10 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
44-811-0 10 mg
44-815-0 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 44-811-0 Supplier Tocris Bioscience™ Supplier No. 4481/10
Add to Cart
Add to Cart
This item is not returnable. View return policy

c-Fms inhibitor; suppresses osteolysis and osteoclast accumulation

Inhibitor of c-Fms tyrosine kinase (M-CSFR, CSF1R) (IC50 values are 2, 12, 217 and 451 nM for c-Fms, VEGFR-2, PDGFRβ and c-Kit respectively). Does not inhibit Flt3, EGFR or c-Src. Suppresses osteoclast differentiation and osteolysis in a rat bone metastasis model.

Chemical Identifiers

CAS 623142-96-1
Molecular Formula C24H24N4O5S

Specifications

Quantity 10 mg
Formula Weight 480.54
Purity >97%
Molecular Weight (g/mol) 480.539
Chemical Name or Material Ki 20227
Recommended Storage Store at +4°C
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.