Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ki-67 Mouse anti-Human, Mouse, Rat, Rhesus, PE, Clone: B56, BD Pharmingen™

Catalog No. BDB567719 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
100 Tests
25 Tests
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB567719 100 Tests
BDB567720 25 Tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. BDB567719 Supplier BD Biosciences Supplier No. 567719
Add to Cart
Add to Cart
This item is not returnable. View return policy

Mouse Monoclonal Antibody

Ki-67 Monoclonal antibody specifically detects Ki-67 in Human, Mouse, Rat, Rhesus samples. It is validated for Intracellular staining (Flow Cytometry)
TRUSTED_SUSTAINABILITY

Specifications

Antigen Ki-67
Applications Flow Cytometry
Classification Monoclonal
Clone B56
Conjugate PE
Dilution 0.02% Sodium Azide
Formulation Aqueous buffered solution containing BSA and ≤0.09% sodium azide.
Gene Alias MKI67; Antigen identified by monoclonal antibody Ki-67; KIA
Host Species Mouse
Immunogen Human Ki-67
Quantity 100 Tests
Regulatory Status RUO
Primary or Secondary Primary
Target Species Human, Mouse, Rat, Rhesus Macaque
Content And Storage Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. The monoclonal antibody was purified from tissue culture supernatant or ascites by affinity chromatography. The antibody was conjugated to the dye under optimum conditions and unconjugated antibody and free dye were removed.
Form Liquid
Isotype IgG1 κ
Show More Show Less
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.