Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ki-67, Mouse Anti-Human, RB613, Clone: B56, BD Horizon™

Catalog No. BDB571126 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
100 Tests
25 Tests
Conjugate:
RB613
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Conjugate
BDB571126 100 Tests RB613
BDB571127 25 Tests RB613
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. BDB571126 Supplier BD Biosciences Supplier No. 571126
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Ki-67 Monoclonal antibody specifically detects Ki-67 in Human (QC Testing), Mouse (Tested in Development), Rat,Dog,Chicken (Reported) samples. It is validated for Flow Cytometry (Intracellular Staining)
TRUSTED_SUSTAINABILITY

Specifications

Antigen Ki-67
Applications Flow Cytometry (Intracellular Staining)
Classification Monoclonal
Clone B56
Conjugate RB613
Formulation Aqueous buffered solution containing ≤0.09% sodium azide.
Gene Alias MKI67; Antigen identified by monoclonal antibody Ki-67; KIA
Host Species Mouse
Immunogen Human Ki-67
Purification Method The monoclonal antibody was purified from tissue culture supernatant or ascites by affinity chromatography.
Quantity 100 Tests
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 4288
Target Species Human, Mouse, Rat, Dog, Chicken
Content And Storage Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze.
Form Purified
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.