Learn More
BD Horizon™ Ki-67 Mouse, anti-Human, RY586, Clone: B56, BD Horizon™
Shop All BD Biosciences ProductsDescription
The B56 monoclonal antibody specifically binds to the Ki-67 antigen that is expressed in the nucleus of cycling cells (G1, S, G2, M cell cycle phases). During the G0 phase, the antigen cannot be detected. During interphase of the cell cycle, it is associated with nucleolar components, and it is on the surface of the chromosomes during M phase. Ki-67 is a large protein having 2 alternatively spliced isoforms, an N-terminal forkhead-associated domain, a C-terminal domain that binds to heterochromatin proteins, and multiple phosphorylation sites, the functions of which are still unclear. Because of the strict association of Ki-67 expression with cell proliferation, anti-Ki-67 antibodies are useful for the identification, quantification, and monitoring of growing cell populations.
Specifications
Specifications
| Antigen | Ki-67 |
| Applications | Flow Cytometry |
| Clone | B56 |
| Conjugate | RY586 |
| Host Species | Mouse |
| Quantity | 25 Tests |
| Regulatory Status | RUO |
| Primary or Secondary | Primary |
| Target Species | Human |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.