Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Ki67/MKI67 Antibody, Alexa Fluor™ 488, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP305537AF488
Description
Ki67/MKI67 Monoclonal antibody specifically detects Ki67/MKI67 in Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/ ImmunofluorescenceSpecifications
Ki67/MKI67 | |
Monoclonal | |
Western Blot, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence | |
antigen identified by monoclonal Ki-67, antigen KI-67, Antigen Ki67, Ki67, Ki-67, KIA, Marker Of Proliferation Ki-67, MIB-, MIB-1, PPP1R105, Proliferation Marker Protein Ki-67, proliferation-related Ki-67 antigen, Protein Phosphatase 1, Regulatory Subunit 105 | |
Recombinant construct containing amino acids 956-1322 of the mouse sequence, Accession Number: XP_006507475, expressed in and purified from E. coli. | |
0.1 mL | |
Breast Cancer, Cancer, Cell Biology, Cell Cycle and Replication, Cellular Markers, Chromatin Research, Mitotic Regulators, Neuroscience | |
4288 | |
Store at 4C in the dark. | |
IgG1 |
Western Blot, Immunohistochemistry, Immunocytochemistry | |
Alexa Fluor 488 | |
50mM Sodium Borate | |
Mouse | |
Immunogen affinity purified | |
RUO | |
Primary | |
Mouse, Rat | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction