Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Ki67/MKI67 Antibody - BSA Free, Novus Biologicals™

Rabbit Polyclonal Antibody has been used in 62 publications
$269.00 - $572.50
Specifications
| Antigen | Ki67/MKI67 |
|---|---|
| Concentration | 1 mg/mL |
| Dilution | Western Blot 1:100-1:2000. Use reported in scientific literature (PMID 22384261), Flow Cytometry 1:100. Use reported by customer review, Immunohistochemistry reported in scientific literature (PMID 28832561), Immunocytochemistry/Immunofluorescence reported in scientific literature (PMID 24779589), Immunohistochemistry-Paraffin 1:100-1:500, Immunohistochemistry-Frozen, Knockout Validated |
| Applications | Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Immunohistochemistry (Frozen), Immunoassay |
| Classification | Polyclonal |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
NB11089717T
![]() |
Novus Biologicals
NB11089717SS |
0.025 mL |
Each for $269.00
|
|
|||||
NB11089717
![]() |
Novus Biologicals
NB11089717 |
0.1 mL |
Each for $572.50
|
|
|||||
Description
Ki67/MKI67 Polyclonal specifically detects Ki67/MKI67 in Human, Mouse, Rat, Porcine samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen, Knockout Validated.Specifications
| Ki67/MKI67 | |
| Western Blot 1:100-1:2000. Use reported in scientific literature (PMID 22384261), Flow Cytometry 1:100. Use reported by customer review, Immunohistochemistry reported in scientific literature (PMID 28832561), Immunocytochemistry/Immunofluorescence reported in scientific literature (PMID 24779589), Immunohistochemistry-Paraffin 1:100-1:500, Immunohistochemistry-Frozen, Knockout Validated | |
| Polyclonal | |
| Rabbit | |
| Breast Cancer, Cancer, Cell Biology, Cell Cycle and Replication, Cellular Markers, Chromatin Research, Mitotic Regulators, Neuroscience | |
| PBS with 0.05% Sodium Azide | |
| antigen identified by monoclonal Ki-67, antigen KI-67, Antigen Ki67, Ki67, Ki-67, KIA, Marker Of Proliferation Ki-67, MIB-, MIB-1, PPP1R105, Proliferation Marker Protein Ki-67, proliferation-related Ki-67 antigen, Protein Phosphatase 1, Regulatory Subunit 105 | |
| MKI67 | |
| IgG | |
| Affinity Purified | |
| Human and mouse. Rat reactivity reported in scientific literature (PMID: 24275061). |
| 1 mg/mL | |
| Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Immunohistochemistry (Frozen), Immunoassay | |
| Unconjugated | |
| RUO | |
| Human, Mouse, Rat, Pig | |
| Q61769 | |
| 4288 | |
| The immunogen for this KI67/MKI67 Antibody was made using a synthetic peptide from the internal region of Mouse KI67/MKI67, between aminoacids 1850-1950 (1899-1916) Uniprot# E9PVX6. | |
| Primary | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. | |
| 351 kDa |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title