Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Ki67/MKI67 Antibody - BSA Free, Novus Biologicals™

Rabbit Polyclonal Antibody has been used in 22 publications
$243.50 - $487.50
Specifications
Antigen | Ki67/MKI67 |
---|---|
Concentration | 1 mg/mL |
Dilution | Western Blot reported in scientific literature (PMID 23643677), Flow Cytometry reported in scientific literature (PMID 25733567), Immunohistochemistry 1:100-1:500, Immunocytochemistry/Immunofluorescence 1:50-1:200, Immunohistochemistry-Paraffin 1:100-1:500, Immunohistochemistry-Frozen reported in scientific literature (PMID 23419702) |
Applications | Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) |
Classification | Polyclonal |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
NB11089719T
![]() |
Novus Biologicals
NB11089719SS |
0.025 mL |
Each for $243.50
|
|
|||||
NB11089719
![]() |
Novus Biologicals
NB11089719 |
0.1 mL |
Each for $487.50
|
|
|||||
Description
Ki67/MKI67 Polyclonal specifically detects Ki67/MKI67 in Human, Mouse, Rat samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen.Specifications
Ki67/MKI67 | |
Western Blot reported in scientific literature (PMID 23643677), Flow Cytometry reported in scientific literature (PMID 25733567), Immunohistochemistry 1:100-1:500, Immunocytochemistry/Immunofluorescence 1:50-1:200, Immunohistochemistry-Paraffin 1:100-1:500, Immunohistochemistry-Frozen reported in scientific literature (PMID 23419702) | |
Polyclonal | |
Rabbit | |
Breast Cancer, Cancer, Cell Biology, Cell Cycle and Replication, Cellular Markers, Chromatin Research, Mitotic Regulators, Neuroscience | |
TBS and 0.1% BSA with 0.1% Sodium Azide | |
antigen identified by monoclonal Ki-67, antigen KI-67, Antigen Ki67, Ki67, Ki-67, KIA, Marker Of Proliferation Ki-67, MIB-, MIB-1, PPP1R105, Proliferation Marker Protein Ki-67, proliferation-related Ki-67 antigen, Protein Phosphatase 1, Regulatory Subunit 105 | |
MKI67 | |
IgG | |
Affinity Purified | |
351 kDa |
1 mg/mL | |
Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
Unconjugated | |
RUO | |
Human, Mouse, Rat | |
P46013 | |
4288 | |
The immunogen for this KI67/MKI67 Antibody was made using a synthetic peptide from the internal region of Mouse KI67/MKI67, between aminoacids 2000-2050 (2021-2036) Uniprot# E9PVX6. | |
Primary | |
Store at 4C. Do not freeze. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title