Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Ki67/MKI67 Antibody (CL1234), Novus Biologicals™

Mouse Monoclonal Antibody
$382.00 - $646.00
Specifications
Antigen | Ki67/MKI67 |
---|---|
Clone | CL1234 |
Dilution | Immunohistochemistry 1:200 - 1:500, Immunocytochemistry/Immunofluorescence 2-10 μg/mL, Immunohistochemistry-Paraffin 1:200 - 1:500 |
Classification | Monoclonal |
Conjugate | Unconjugated |
Description
Ki67/MKI67 Monoclonal specifically detects Ki67/MKI67 in Human samples. It is validated for Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.Specifications
Ki67/MKI67 | |
Immunohistochemistry 1:200 - 1:500, Immunocytochemistry/Immunofluorescence 2-10 μg/mL, Immunohistochemistry-Paraffin 1:200 - 1:500 | |
Unconjugated | |
Mouse | |
PBS, pH 7.2, containing 40% glycerol with 0.02% sodium azide | |
4288.0 | |
This KI67/MKI67 Antibody was developed against Recombinant Protein corresponding to amino acids 1-159 from Human KI67/MKI67: DGPHFPLSLSTCLFGRGIECDIRIQLPVVSKQHCKIEIHEQEAILHNFSSTNPTQVNGSVIDEPVRLKHGDVITIIDRSFRYENESLQSGRKSTEFPRKIREQEPARRVSRSSFSSDPDEKAQDSKAYSK | |
Primary | |
Specificity of Ki67/MKI67 Antibody (CL1234) verified on a Protein Array containing target protein plus 383 other non-specific proteins. |
CL1234 | |
Monoclonal | |
Purified | |
Breast Cancer, Cancer, Cell Biology, Cell Cycle and Replication, Cellular Markers, Chromatin Research, Mitotic Regulators, Neuroscience | |
antigen identified by monoclonal Ki-67, antigen KI-67, Antigen Ki67, Ki67, Ki-67, KIA, Marker Of Proliferation Ki-67, MIB-, MIB-1, PPP1R105, Proliferation Marker Protein Ki-67, proliferation-related Ki-67 antigen, Protein Phosphatase 1, Regulatory Subunit 105 | |
MKI67 | |
IgG1 | |
Protein A purified | |
359 kDa |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title