Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Ki67/MKI67 Antibody, mFluor Violet 450 SE, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NB11089717MFV450
Description
Ki67/MKI67 Polyclonal antibody specifically detects Ki67/MKI67 in Human, Mouse, Rat, Porcine samples. It is validated for Flow CytometrySpecifications
Ki67/MKI67 | |
Polyclonal | |
Flow Cytometry | |
antigen identified by monoclonal Ki-67, antigen KI-67, Antigen Ki67, Ki67, Ki-67, KIA, Marker Of Proliferation Ki-67, MIB-, MIB-1, PPP1R105, Proliferation Marker Protein Ki-67, proliferation-related Ki-67 antigen, Protein Phosphatase 1, Regulatory Subunit 105 | |
The immunogen for this KI67/MKI67 Antibody was made using a synthetic peptide from the internal region of Mouse KI67/MKI67, between aminoacids 1850-1950 (1899-1916) Uniprot# E9PVX6. | |
0.1 mL | |
Breast Cancer, Cancer, Cell Biology, Cell Cycle and Replication, Cellular Markers, Chromatin Research, Mitotic Regulators, Neuroscience | |
4288 | |
Store at 4C in the dark. | |
IgG |
Flow Cytometry | |
mFluor Violet 450 SE | |
50mM Sodium Borate | |
Rabbit | |
Immunogen affinity purified | |
RUO | |
Primary | |
Human, Mouse, Rat, Pig | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction