Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ KIAA0020 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA531247
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA531247 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA531247 Supplier Invitrogen™ Supplier No. PA531247
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Recommended positive controls: 293T, A431, Jurkat, Raji. Predicted reactivity: Mouse (87%), Rat (86%), Bovine (89%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.

Inhibits the poly(ADP-ribosyl)ation activity of PARP1 and the degradation of PARP1 by CASP3 following genotoxic stress. Binds to double-stranded RNA or DNA without sequence specificity. Involved in development of the eye and of primordial germ cells.
TRUSTED_SUSTAINABILITY

Specifications

Antigen KIAA0020
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 20% glycerol and 0.01% thimerosal; pH 7
Gene PUM3
Gene Accession No. Q15397
Gene Alias cPERP-C; HA-8; HBV XAg-transactivated protein 5; HBV X-transactivated gene 5 protein; HLA-HA8; KIAA0020; Minor histocompatibility antigen HA-8; PEN; penguin homolog; protein 5 transactivated by hepatitis B virus X antigen (HBxAg); PUF6; PUF-A; PUM3; Pumilio homolog 3; pumilio RNA binding family member 3; pumilio RNA-binding family member 3; XTP5
Gene Symbols PUM3
Host Species Rabbit
Immunogen Recombinant protein encompassing a sequence within the C-terminus region of human KIAA0020. The exact sequence is proprietary.
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 9933
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.