Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ KIAA0040 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA564386
Description
Immunogen sequence: KKKKKDEEDL WISAQPKLLQ MEKRPSLPV Highest antigen sequence identity to the following orthologs: Mouse - 90%, Rat - 45%.
The specific function of KIAA0040 is not yet known.
Specifications
KIAA0040 | |
Polyclonal | |
Unconjugated | |
KIAA0040 | |
k0040; KIAA0040; uncharacterized protein KIAA0040 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
9674 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
0.1 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q15053 | |
KIAA0040 | |
Recombinant protein corresponding to Human KIAA0040. Recombinant protein control fragment (Product #RP-103766). | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction