Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ KIAA0040 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA541556
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA541556 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA541556 Supplier Invitrogen™ Supplier No. PA541556
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Peptide sequence: DPKVALESLY PLAKFTAELG LSPDKAAEVL SQGAHLAAGP DGRTIDRFSP Sequence homology: Human: 100%.

The specific function of KIAA0040 is not yet known.
TRUSTED_SUSTAINABILITY

Specifications

Antigen KIAA0040
Applications Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 2% sucrose and 0.09% sodium azide
Gene KIAA0040
Gene Accession No. Q15053
Gene Alias k0040; KIAA0040; uncharacterized protein KIAA0040
Gene Symbols KIAA0040
Host Species Rabbit
Immunogen Synthetic peptide directed towards the middle region of human KIAA0040.
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 9674
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.