Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ KIAA0087 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA599749
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA599749 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA599749 Supplier Invitrogen™ Supplier No. PA599749
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Antibody detects endogenous levels of total KIAA0087.

KIAA0087 (KIAA0087 LncRNA) is an RNA Gene, and is affiliated with the non-coding RNA class. Diseases associated with KIAA0087 include Endometrial Cancer and Pancreatic Cancer.
TRUSTED_SUSTAINABILITY

Specifications

Antigen KIAA0087
Applications Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Gene KIAA0087
Gene Accession No. Q14695
Gene Alias KIAA0087
Gene Symbols KIAA0087
Host Species Rabbit
Immunogen A synthesized peptide derived from human KIAA0087(Accession Q14695), corresponding to amino acid residues N48-N98.
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 9808
Target Species Human, Rat
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.