Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ KIAA0090 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA561834
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA561834 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA561834 Supplier Invitrogen™ Supplier No. PA561834
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: LKKTTLALHH LSSGHLKWVE HLPESDSIHY QMVYSYGSGV VWALGVVPFS HVNIVKFNVE DGEIVQQVRV STPWLQHLSG ACGVVDEAVL VCPDPSSRSL QTL Highest antigen sequence identity to the following orthologs: Mouse - 93%, Rat - 93%.

This gene encodes a single-pass type I transmembrane protein, which is a subunit of the endoplasmic reticulum membrane protein complex (EMC). Multiple alternatively spliced transcript variants encoding different isoforms have been found for this gene.
TRUSTED_SUSTAINABILITY

Specifications

Antigen KIAA0090
Applications Immunohistochemistry (Paraffin), Immunocytochemistry
Classification Polyclonal
Concentration 0.1 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene EMC1
Gene Accession No. Q8N766
Gene Alias 2700016F22Rik; CAVIPMR; Emc1; ER membrane protein complex subunit 1; KIAA0090; novel DUF1620 domain containing protein; PSEC0263; RGD1310427; RP1-43E13.1
Gene Symbols EMC1
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human KIAA0090. Recombinant protein control fragment (Product #RP-99795).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 23065
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.