Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ KIAA0100 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA560118
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA560118 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA560118 Supplier Invitrogen™ Supplier No. PA560118
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: EHPVDDIDKM KERAAMNNSF IYIKIPQVPL CVSYKGEKNS VDWGDLNLVL PCLEYHNNTW TWLDFAMAVK RDSRKALVAQ VIKEKLRLKS ATGSEVRGKL ETKSDLN Highest antigen sequence identity to the following orthologs: Mouse - 98%, Rat - 98%.

Belongs to the UPF0378 family. 4 isoforms of the human protein are produced by alternative splicing. Song J., Biochim. Biophys. Acta 1760:62-69(2006). Nagase T., DNA Res. 2:37-43(1995). Chen G., Leuk. Res. 29:503-509(2005).
TRUSTED_SUSTAINABILITY

Specifications

Antigen KIAA0100
Applications Immunohistochemistry (Paraffin), Immunocytochemistry
Classification Polyclonal
Concentration 0.2 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene KIAA0100
Gene Accession No. Q14667
Gene Alias Antigen MLAA-22; BCOX; BCOX1; breast cancer overexpressed gene 1; breast cancer-overexpressed gene 1 protein; cancer/testis antigen 101; CT101; FMP27; K0100; KIAA0100; protein KIAA0100; U937-associated antigen
Gene Symbols KIAA0100
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human KIAA0100. Recombinant protein control fragment (Product #RP-100185).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 9703
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.