Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ KIAA0101 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA565485
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA565485 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA565485 Supplier Invitrogen™ Supplier No. PA565485
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: VRTKADSVPGT YRKVVAARAP RKVLGSSTSA TNSTSVSSRK Highest antigen sequence identity to the following orthologs - mouse 76%, rat 78%.

PCNA-binding protein that acts as a regulator of DNA repair during DNA replication. Following DNA damage, the interaction with PCNA is disrupted, facilitating the interaction between monoubiquitinated PCNA and the translesion DNA synthesis DNA polymerase eta (POLH) at stalled replisomes, facilitating the bypass of replication-fork-blocking lesions. Also acts as a regulator of centrosome number.
TRUSTED_SUSTAINABILITY

Specifications

Antigen KIAA0101
Applications Immunocytochemistry
Classification Polyclonal
Concentration 0.05 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene PCLAF
Gene Accession No. Q15004
Gene Alias 2810417H13Rik; AA409629; HCV NS5A-transactivated protein 9; HCV NS5A-transactivated protein 9 homolog; hepatitis C virus NS5A-transactivated protein 9; hypothetical protein LOC540737; KIAA0101; L5; mKIAA0101; NS5A (hepatitis C virus) transactivated protein 9; Ns5apt9; NS5ATP9; OEATC; OEATC1; OEATC-1; overexpressed in anaplastic thyroid carcinoma 1; p15(PAF); p15/PAF; p15PAF; PAF; PAF15; Pclaf; PCNA clamp associated factor; PCNA-associated factor; PCNA-associated factor of 15 kDa; PCNA-associated factor-like protein; PCNA-clamp-associated factor; RIKEN cDNA 2810417H13 gene
Gene Symbols PCLAF
Host Species Rabbit
Immunogen Recombinant Human KIAA0101. Recombinant protein control fragment (Product #RP-106159).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 9768
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.