Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ KIAA0196 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA567132
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA567132 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA567132 Supplier Invitrogen™ Supplier No. PA567132
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: HRGLIFNPRAK PSELMPKLKE LGATMDGFHR SFEYIQDYVN IYGLKIWQEE VSRIINYNVE QECNNFLRTK IQDWQSMYQS THIPIPKFTP VD Highest antigen sequence identity to the following orthologs - mouse 98%, rat 98%.

This gene encodes a 134 kDa protein named strumpellin that is predicted to have multiple transmembrane domains and a spectrin-repeat-containing domain. This ubiquitously expressed gene has its highest expression in skeletal muscle. The protein is named for Strumpell disease; a form of hereditary spastic paraplegia (HSP). Spastic paraplegias are a diverse group of disorders in which the autosomal dominant forms are characterized by progressive, lower extremity spasticity caused by axonal degeneration in the terminal portions of the longest descending and ascending corticospinal tracts. More than 30 loci (SPG1-33) have been implicated in hereditary spastic paraplegia diseases.
TRUSTED_SUSTAINABILITY

Specifications

Antigen KIAA0196
Applications Immunocytochemistry
Classification Polyclonal
Concentration 0.1 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene WASHC5
Gene Accession No. Q12768
Gene Alias AL022848; C76463; E430025E21Rik; KIAA0196; mKIAA0196; RGD1564420; RIKEN cDNA E430025E21 gene; RTSC; RTSC1; similar to Hypothetical protein MGC31278; SPG8; Strumpellin; uncharacterized protein LOC362919; WASH complex subunit 5; WASH complex subunit strumpellin; WASHC5
Gene Symbols WASHC5
Host Species Rabbit
Immunogen Recombinant Human KIAA0196. Recombinant protein control fragment (Product #RP-107779).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 9897
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.