Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ KIAA0247 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA556481
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA556481 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA556481 Supplier Invitrogen™ Supplier No. PA556481
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: NGGYICHPRP CRDPLTAGSV IEYLCAEGYM LKGDYKYLTC KNGEWKPAME ISCRLNEDKD THTSLGVPTL S Highest antigen sequence identity to the following orthologs: Mouse - 87%, Rat - 90%.

KIAA0247 is a single-pass type I membrane protein. It contains 1 Sushi (CCP/SCR) domain. The function of the KIAA0247 protein remains unknown.
TRUSTED_SUSTAINABILITY

Specifications

Antigen KIAA0247
Applications Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 0.50 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene SUSD6
Gene Accession No. Q92537
Gene Alias 4933426M11Rik; DRAGO; drug-activated gene overexpressed protein; Kiaa0247; LOC678893; mKIAA0247; similar to Protein KIAA0247 precursor; SUSD6; sushi domain containing 6; sushi domain-containing protein 6
Gene Symbols SUSD6
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human KIAA0247. Recombinant protein control fragment (Product #RP-89126).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 9766
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.